4-Brom-3,5-xylenol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Brom-5-hydroxy-3-xylol, 4-Brom-3,5-dimethylphenol
OT-0433-5G
OT-0433-25G
OT-0433-100G
COBBOT-0433-5GEA
37.9
EUR
COBBOT-0433-5G
COBBOT-0433-25G
COBBOT-0433-100G
4-Brom-3,5-xylenol
4-Brom-3,5-xylenol
Formel:
C₈H₉BrO Molecular Weight: 201,06 g/mol Siedepunkt: 259 °C (1013 hPa) Schmelzpunkt: 116 °C |
MDL:
MFCD00002315 CAS-Nummer: 7463-51-6 EINECS: 231-255-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...