1-Brom-2-chlor-3-fluorbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-3-fluorobromobenzene

OT-0550-100G OT-0550-25G OT-0550-5G
COBBOT-0550-100GEA 460 EUR
COBBOT-0550-100G COBBOT-0550-25G COBBOT-0550-5G
1-Brom-2-chlor-3-fluorbenzol
1-Bromo-2-chloro-3-fluorobenzene
Formel: C₆H₃BrClF
Molecular Weight: 209.44 g/mol
MDL: MFCD04038306
CAS-Nummer: 883499-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR