1-Brom-2-chlor-4-fluorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
4-Brom-3-chlor-1-fluorbenzol
005539-1G
005539-5G
005539-25G
FLUO005539-1GEA
31.1
EUR
FLUO005539-1G
FLUO005539-5G
FLUO005539-25G
1-Brom-2-chlor-4-fluorbenzol
1-Bromo-2-chloro-4-fluorobenzene
Formel:
C₆H₃BrClF Molecular Weight: 209.44 g/mol |
MDL:
MFCD00142587 CAS-Nummer: 110407-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...