4-Brom-3-chloranisol

Lieferant: COMBI-BLOCKS INC.

COBBOT-0446-5GEA 49 EUR
COBBOT-0446-5G COBBOT-0446-25G COBBOT-0446-100G
4-Brom-3-chloranisol
4-Brom-3-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
MDL: MFCD00070742
CAS-Nummer: 50638-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR