1-Brom-2-chlor-5-fluorbenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Bromo-1-chloro-4-fluorobenzene
OT-0491-25G
OT-0491-100G
OT-0491-5G
COBBOT-0491-25GEA
63.9
EUR
COBBOT-0491-25G
COBBOT-0491-100G
COBBOT-0491-5G
1-Brom-2-chlor-5-fluorbenzol
1-Brom-2-chlor-5-fluorbenzol
Formel:
C₆H₃BrClF Molecular Weight: 209,44 g/mol |
MDL:
MFCD00061120 CAS-Nummer: 201849-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...