2-Brom-4-fluorbenzonitril

Lieferant: CHEMPUR

05-0062.1 05-0062.5
CHMP05-0062.1EA 0 EUR
CHMP05-0062.1 CHMP05-0062.5
2-Brom-4-fluorbenzonitril
2-Brom-4-fluorbenzonitril
Formel: C₇H₃BrFN
Molecular Weight: 200,01 g/mol
MDL: MFCD00672924
CAS-Nummer: 36282-26-5
EINECS: 252-949-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR