3-Brom-2-fluorphenol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-2-fluor-3-hydroxybenzol

OT-0701-5G OT-0701-1G
COBBOT-0701-5GEA 493 EUR
COBBOT-0701-5G COBBOT-0701-1G
3-Brom-2-fluorphenol
3-Brom-2-fluorphenol
Formel: C₆H₄BrFO
MDL: MFCD09864673
CAS-Nummer: 156682-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR