1-Brom-2-fluor-3-nitrobenzol
Lieferant: COMBI-BLOCKS
Synonyme:
3-Brom-2-fluornitrobenzol
OS-7453-1G
OS-7453-5G
COBBOS-7453-1GEA
61.3
EUR
COBBOS-7453-1G
COBBOS-7453-5G
1-Brom-2-fluor-3-nitrobenzol
1-Bromo-2-fluoro-3-nitrobenzene
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol |
MDL:
MFCD05663754 CAS-Nummer: 58534-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...