1-Brom-2-fluor-4-iod-5-nitrobenzol

Lieferant: COMBI-BLOCKS

HC-3154-5G HC-3154-1G
COBBHC-3154-5GEA 884 EUR
COBBHC-3154-5G COBBHC-3154-1G
1-Brom-2-fluor-4-iod-5-nitrobenzol
1-Bromo-2-fluoro-4-iodo-5-nitrobenzene
Formel: C₆H₂BrFINO₂
Molecular Weight: 345,89 g/mol
MDL: MFCD16295088
CAS-Nummer: 1226808-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR