4-Brom-3-fluortoluol
Lieferant: COMBI-BLOCKS
|
Gefahr
|
Synonyme:
1-Bromo-2-fluoro-4-methylbenzene
AN-1047-25G
AN-1047-100G
COBBAN-1047-25GEA
42.3
EUR
COBBAN-1047-25G
COBBAN-1047-100G
4-Brom-3-fluortoluol
4-Brom-3-fluortoluol
Formel:
C₇H₆BrF Molecular Weight: 189.03 g/mol Siedepunkt: 186 Dichte: 1.49 g/cm³ (20 °C) |
MDL:
MFCD00040940 CAS-Nummer: 452-74-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...