1-Brom-2-fluor-4-nitrobenzol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-3-fluornitrobenzol
OS-7449-5G
OS-7449-25G
OS-7449-100G
COBBOS-7449-5GEA
31
EUR
COBBOS-7449-5G
COBBOS-7449-25G
COBBOS-7449-100G
1-Brom-2-fluor-4-nitrobenzol
1-Bromo-2-fluoro-4-nitrobenzene
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol |
MDL:
MFCD05865088 CAS-Nummer: 185331-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...