1-Brom-2-fluor-5-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-1-fluor-4-nitrobenzol, 3-Brom-4-fluornitrobenzol

OS-7447-5G OS-7447-25G OS-7447-100G
COBBOS-7447-5GEA 26.2 EUR
COBBOS-7447-5G COBBOS-7447-25G COBBOS-7447-100G
1-Brom-2-fluor-5-nitrobenzol
1-Brom-2-fluor-5-nitrobenzol
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD00055433
CAS-Nummer: 701-45-1
EINECS: 211-855-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR