1-Brom-2-fluor-5-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-1-fluor-4-nitrobenzol, 3-Brom-4-fluornitrobenzol

021887-25G 021887-5G
FLUO021887-25GEA 33.6 EUR
FLUO021887-25G FLUO021887-5G
1-Brom-2-fluor-5-nitrobenzol
1-Brom-2-fluor-5-nitrobenzol
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD00055433
CAS-Nummer: 701-45-1
EINECS: 211-855-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR