1-Brom-2-fluor-6-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-1-fluor-3-nitrobenzol, 2-Brom-3-fluornitrobenzol

OS-7445-5G OS-7445-25G OS-7445-1G
COBBOS-7445-5GEA 260 EUR
COBBOS-7445-5G COBBOS-7445-25G COBBOS-7445-1G
1-Brom-2-fluor-6-nitrobenzol
1-Bromo-2-fluoro-6-nitrobenzene
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD07782080
CAS-Nummer: 59255-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR