2-Brom-5-methoxyphenol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-2-hydroxy-4-methoxybenzol, 4-Brom-3-hydroxyanisol

OT-0669-5G OT-0669-1G
COBBOT-0669-5GEA 312 EUR
COBBOT-0669-5G COBBOT-0669-1G
2-Brom-5-methoxyphenol
2-Brom-5-methoxyphenol
Formel: C₇H₇BrO₂
MDL: MFCD03428523
CAS-Nummer: 63604-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR