2-Brom-4-(trifluormethyl)phenol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-4-hydroxybenzotrifluoride

OT-1249-5G OT-1249-25G OT-1249-1G
COBBOT-1249-5GEA 76.8 EUR
COBBOT-1249-5G COBBOT-1249-25G COBBOT-1249-1G
2-Brom-4-(trifluormethyl)phenol
2-Brom-4-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
Molecular Weight: 241.01
MDL: MFCD06657954
CAS-Nummer: 81107-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR