2-Brom-4-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

Synonyme: 3-Bromo-4-hydroxybenzotrifluoride

023859-25G 023859-5G
FLUO023859-25GEA 230 EUR
FLUO023859-25G FLUO023859-5G
2-Brom-4-(trifluormethyl)phenol
2-Brom-4-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
Molecular Weight: 241.01
MDL: MFCD06657954
CAS-Nummer: 81107-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR