2-Brom-4-methoxyphenol
Lieferant: BIONET RESEARCH
Synonyme:
1-Brom-2-hydroxy-5-methoxybenzol, 3-Brom-4-hydroxyanisol
5Y-0804.500
5Y-0804.5
5Y-0804.10
5Y-0804.1
BIOT5Y-0804.500EA
0
EUR
BIOT5Y-0804.500
BIOT5Y-0804.5
BIOT5Y-0804.10
BIOT5Y-0804.1
2-Brom-4-methoxyphenol
2-Brom-4-methoxyphenol
Formel:
C₇H₇BrO₂ Molecular Weight: 203.04 g/mol |
MDL:
MFCD02113712 CAS-Nummer: 17332-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...