2-Brom-4-methoxyphenol
Lieferant: COMBI-BLOCKS
Synonyme:
1-Brom-2-hydroxy-5-methoxybenzol, 3-Brom-4-hydroxyanisol
OT-0487-100G
OT-0487-5G
OT-0487-1G
OT-0487-25G
COBBOT-0487-100GEA
1050
EUR
COBBOT-0487-100G
COBBOT-0487-5G
COBBOT-0487-1G
COBBOT-0487-25G
2-Brom-4-methoxyphenol
2-Brom-4-methoxyphenol
Formel:
C₇H₇BrO₂ Molecular Weight: 203.04 g/mol |
MDL:
MFCD02113712 CAS-Nummer: 17332-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...