4-Brom-3-iodbenzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-5-(trifluormethyl)iodbenzol, 4-Brom-3-iodbenzotrifluorid, 1-Brom-2-iod-4-(trifluormethyl)benzol

HC-3222-5G HC-3222-25G HC-3222-1G
COBBHC-3222-5GEA 114 EUR
COBBHC-3222-5G COBBHC-3222-25G COBBHC-3222-1G
4-Brom-3-iodbenzotrifluorid
4-Brom-3-iodbenzotrifluorid
Formel: C₇H₃BrF₃I
Molecular Weight: 350,9 g/mol
MDL: MFCD09800691
CAS-Nummer: 640280-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR