1-Brom-2-iodbenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
HC-2601-25G
HC-2601-100G
COBBHC-2601-25GEA
50.9
EUR
COBBHC-2601-25G
COBBHC-2601-100G
1-Brom-2-iodbenzol
1-Brom-2-iodbenzol
Formel:
C₆H₄BrI Molecular Weight: 282.91 g/mol Siedepunkt: 250 Dichte: 2.2571 g/cm³ (25 °C) |
MDL:
MFCD00001030 CAS-Nummer: 583-55-1 EINECS: 209-508-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...