2-(3-Bromphenyl)ethanol

Lieferant: COMBI-BLOCKS

OT-0409-5G OT-0409-25G OT-0409-1G
COBBOT-0409-5GEA 50.9 EUR
COBBOT-0409-5G COBBOT-0409-25G COBBOT-0409-1G
2-(3-Bromphenyl)ethanol
2-(3-Bromphenyl)ethanol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00191857
CAS-Nummer: 28229-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR