3-tert-Butyl-1-brombenzol

Lieferant: FLUOROCHEM

Synonyme: 2-(3-Bromphenyl)-2-methylpropan, 3-Brom-tert-butylbenzol, 1-Brom-3-tert-butylbenzol

023850-25G 023850-5G
FLUO023850-25GEA 248 EUR
FLUO023850-25G FLUO023850-5G
3-tert-Butyl-1-brombenzol
3-tert-Butyl-1-brombenzol
Formel: C₁₀H₁₃Br
Molecular Weight: 213.12 g/mol
MDL: MFCD00234661
CAS-Nummer: 3972-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR