3-Brombenzylalkohol

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromophenyl)methanol

OS-7638-5G OS-7638-25G OS-7638-100G
COBBOS-7638-5GEA 26.2 EUR
COBBOS-7638-5G COBBOS-7638-25G COBBOS-7638-100G
3-Brombenzylalkohol
3-Brombenzylalkohol
Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Siedepunkt: 165 °C (1013 hPa)
Dichte: 1,56 g/cm³ (20 °C)
MDL: MFCD00004629
CAS-Nummer: 15852-73-0
EINECS: 239-975-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR