3-Brombenzophenon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
m-Brombenzophenon
H53505.03
H53505.06
H53505.14
H53505.03EA
88.4
EUR
H53505.03
H53505.06
H53505.14
3-Brombenzophenon ≥97%
3-Brombenzophenon
Purity: 97%
CAS No.: 1016-77-9
Molecular Formula: C13H9BrO
Formula Weight: 261.12
Melting Point: 74-78°
MDL No.: MFCD00672015
Formel:
C₁₃H₉BrO Molecular Weight: 261,12 g/mol Siedepunkt: 355 °C (1013 hPa) Schmelzpunkt: 74…78 °C Dichte: 1,42 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672015 CAS-Nummer: 1016-77-9 EINECS: 213-808-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...