2-Brom-2,6-dichlortoluol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-3,5-dichlor-2-methylbenzol

OT-0578-1G OT-0578-5G
COBBOT-0578-1GEA 303 EUR
COBBOT-0578-1G COBBOT-0578-5G
2-Brom-2,6-dichlortoluol
2-Brom-2,6-dichlortoluol
Formel: C₇H₅BrCl₂
Molecular Weight: 239,93 g/mol
MDL: MFCD09801039
CAS-Nummer: 115615-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR