2-Brom-4,6-difluorphenol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
1-Bromo-3,5-difluoro-2-hydroxybenzene
OT-0949-25G
OT-0949-5G
OT-0949-100G
COBBOT-0949-25GEA
163
EUR
COBBOT-0949-25G
COBBOT-0949-5G
COBBOT-0949-100G
2-Brom-4,6-difluorphenol
2-Brom-4,6-difluorphenol
Formel:
C₆H₃BrF₂O Molecular Weight: 208,99 g/mol |
MDL:
MFCD00070752 CAS-Nummer: 98130-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...