2-Brom-6-chloranisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-3-chloro-2-methoxybenzene

OT-1144-5G OT-1144-25G OT-1144-1G
COBBOT-1144-5GEA 629 EUR
COBBOT-1144-5G COBBOT-1144-25G COBBOT-1144-1G
2-Brom-6-chloranisol
2-Brom-6-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
MDL: MFCD15143586
CAS-Nummer: 174913-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR