4-Brom-2-chlorbenzonitril 96%

Lieferant: Thermo Scientific
Gefahr

L10602.03 L10602.06
L10602.03EA 16.4 EUR
L10602.03 L10602.06
4-Brom-2-chlorbenzonitril 96%
4-Brom-2-chlorbenzonitril

Purity: 96% CAS No.: 154607-01-9 Molecular Formula: C7H3BrClN Formula Weight: 216.47 Melting Point: 67-72° Boiling Point: 142-143°/10mm UN No.: 3439 MDL No.: MFCD00040883

Formel: C₇H₃BrClN
Molecular Weight: 216,46 g/mol
Siedepunkt: 142…143 °C (10 mmHg)
Schmelzpunkt: 67…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040883
CAS-Nummer: 154607-01-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR