4-Brom-2-chlor-6-fluorphenol

Lieferant: COMBI-BLOCKS

OT-1146-1G OT-1146-5G
COBBOT-1146-1GEA 0 EUR
COBBOT-1146-1G COBBOT-1146-5G
4-Brom-2-chlor-6-fluorphenol
4-Brom-2-chlor-6-fluorphenol
Formel: C₆H₃BrClFO
Molecular Weight: 225,44 g/mol
MDL: MFCD01631476
CAS-Nummer: 161045-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR