3-Brom-5-chloranisol
Lieferant: COMBI-BLOCKS
Synonyme:
1-Bromo-3-chloro-5-methoxybenzene
OT-0786-5G
OT-0786-1G
COBBOT-0786-5GEA
107
EUR
COBBOT-0786-5G
COBBOT-0786-1G
3-Brom-5-chloranisol
3-Brom-5-chloranisol
Formel:
C₇H₆BrClO Molecular Weight: 221,48 g/mol |
MDL:
MFCD11035921 CAS-Nummer: 174913-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...