4-Brom-2-chlor-5-fluoranisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-5-chlor-2-fluor-4-methoxybenzol

OT-1213-5G OT-1213-1G OT-1213-250M
COBBOT-1213-5GEA 1810 EUR
COBBOT-1213-5G COBBOT-1213-1G COBBOT-1213-250M
4-Brom-2-chlor-5-fluoranisol
4-Brom-2-chlor-5-fluoranisol
Formel: C₇H₅BrClFO
Molecular Weight: 239,47 g/mol
MDL: MFCD16036128
CAS-Nummer: 901236-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR