2-Brom-4-chlor-1-fluorbenzol

Lieferant: CHEMPUR

Synonyme: 3-Brom-1-chlor-4-fluorbenzol, 1-Brom-3-chlor-6-fluorbenzol

FL13192.POR 013192-25G
CHMPFL13192.POREA 0 EUR
CHMPFL13192.POR FLUO013192-25G
2-Brom-4-chlor-1-fluorbenzol
2-Brom-4-chlor-1-fluorbenzol
Formel: C₆H₃BrClF
Molecular Weight: 209.44 g/mol
MDL: MFCD00672934
CAS-Nummer: 1996-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR