2-Brom-4-chlor-1-fluorbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-1-chlor-4-fluorbenzol, 1-Brom-3-chlor-6-fluorbenzol

M-Clarity™ Qualitätsstufe = MQ100

548685-25G 548685-5G
SIAL548685-25GEA 85 EUR
SIAL548685-25G SIAL548685-5G
2-Brom-4-chlor-1-fluorbenzol, Sigma-Aldrich®
2-Brom-4-chlor-1-fluorbenzol
Formel: C₆H₃BrClF
Molecular Weight: 209,44 g/mol
MDL: MFCD00672934
CAS-Nummer: 1996-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR