2-Brom-4-chloranisol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-4-chloro-1-methoxybenzene
OT-0860-25G
OT-0860-100G
COBBOT-0860-25GEA
51.3
EUR
COBBOT-0860-25G
COBBOT-0860-100G
2-Brom-4-chloranisol
2-Brom-4-chloranisol
Formel:
C₇H₆BrClO Molecular Weight: 221,48 g/mol |
MDL:
MFCD00079705 CAS-Nummer: 60633-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...