3-Brom-2-fluorbenzonitril

Lieferant: FLUOROCHEM

023918-1G 023918-5G
FLUO023918-1GEA 31.1 EUR
FLUO023918-1G FLUO023918-5G
3-Brom-2-fluorbenzonitril
3-Brom-2-fluorbenzonitril
Formel: C₇H₃BrFN
Molecular Weight: 200.01 g/mol
MDL: MFCD06657980
CAS-Nummer: 840481-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR