5-Brom-2-methoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-2-methoxybenzoesäurenitril

M-Clarity™ Qualitätsstufe = MQ100

701173-25G 701173-5G
SIAL701173-25GEA 421 EUR
SIAL701173-25G SIAL701173-5G
5-Brom-2-methoxybenzonitril, Sigma-Aldrich®
5-Brom-2-methoxybenzonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD00143429
CAS-Nummer: 144649-99-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR