3-Brom-4-methoxybenzonitril ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Brom-4-methoxybenzoesäurenitril

A16698.06 A16698.14
A16698.06EA 32.9 EUR
A16698.06 A16698.14
3-Brom-4-methoxybenzonitril ≥99%
3-Brom-4-methoxybenzonitril

Purity: 99% CAS No.: 117572-79-9 Molecular Formula: C8H6BrNO Formula Weight: 212.05 Melting Point: 120-124° Beilstein Reference: 3246576 UN No.: 3439 MDL No.: MFCD00143097

Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143097
CAS-Nummer: 117572-79-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR