3-Brom-4-methoxybenzonitril

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-4-methoxybenzoesäurenitril

CA-4624-5G CA-4624-25G CA-4624-1G CA-4624-100G
COBBCA-4624-5GEA 61.9 EUR
COBBCA-4624-5G COBBCA-4624-25G COBBCA-4624-1G COBBCA-4624-100G
3-Brom-4-methoxybenzonitril
3-Brom-4-methoxybenzonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD00143097
CAS-Nummer: 117572-79-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR