2-Brom-6-fluoranisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-3-fluoro-2-methoxybenzene

OT-0447-25G OT-0447-5G
COBBOT-0447-25GEA 364 EUR
COBBOT-0447-25G COBBOT-0447-5G
2-Brom-6-fluoranisol
2-Brom-6-fluoranisol
Formel: C₇H₆BrFO
Molecular Weight: 205.03 g/mol
MDL: MFCD04974125
CAS-Nummer: 845829-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR