1-Brom-5-fluor-4-iod-2-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-3-fluor-4-iod-6-nitrobenzol

AN-1532-5G AN-1532-1G
COBBAN-1532-5GEA 595 EUR
COBBAN-1532-5G COBBAN-1532-1G
1-Brom-5-fluor-4-iod-2-nitrobenzol
1-Brom-5-fluor-4-iod-2-nitrobenzol
Formel: C₆H₂BrFINO₂
Molecular Weight: 345,89 g/mol
MDL: MFCD12913932
CAS-Nummer: 1187385-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR