1-Brom-3-fluor-4-iodbenzol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 4-Brom-2-fluor-1-iodbenzol

HC-3047-25G HC-3047-100G
COBBHC-3047-25GEA 48.1 EUR
COBBHC-3047-25G COBBHC-3047-100G
1-Brom-3-fluor-4-iodbenzol
1-Brom-3-fluor-4-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
Siedepunkt: 241 °C (1013 hPa)
Schmelzpunkt: 46…49 °C
MDL: MFCD00010608
CAS-Nummer: 105931-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR