1-Brom-3-fluor-4-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-2-fluornitrobenzol

HC-6681-25G HC-6681-5G HC-6681-1G
COBBHC-6681-25GEA 127 EUR
COBBHC-6681-25G COBBHC-6681-5G COBBHC-6681-1G
1-Brom-3-fluor-4-nitrobenzol
1-Bromo-3-fluoro-4-nitrobenzene
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD01930221
CAS-Nummer: 321-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR