2-Brom-4-fluor-1-iodbenzol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 1-Brom-5-fluor-2-iodbenzol, 3-Brom-1-fluor-4-iodbenzol

HC-2956-5G HC-2956-25G HC-2956-100G
COBBHC-2956-5GEA 28.5 EUR
COBBHC-2956-5G COBBHC-2956-25G COBBHC-2956-100G
2-Brom-4-fluor-1-iodbenzol
2-Brom-4-fluor-1-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
MDL: MFCD00142868
CAS-Nummer: 202865-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR