1-Brom-5-fluor-2-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-4-fluornitrobenzol, 1-Brom-3-fluor-6-nitrobenzol

OS-7443-5G OS-7443-25G OS-7443-1G
COBBOS-7443-5GEA 51.3 EUR
COBBOS-7443-5G COBBOS-7443-25G COBBOS-7443-1G
1-Brom-5-fluor-2-nitrobenzol
1-Brom-5-fluor-2-nitrobenzol
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD00792441
CAS-Nummer: 700-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR