1-Brom-5-fluor-2-nitrobenzol
Lieferant: FLUOROCHEM
Synonyme:
2-Brom-4-fluornitrobenzol, 1-Brom-3-fluor-6-nitrobenzol
008649-1G
008649-5G
FLUO008649-1GEA
0
EUR
FLUO008649-1G
FLUO008649-5G
1-Brom-5-fluor-2-nitrobenzol
1-Brom-5-fluor-2-nitrobenzol
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol |
MDL:
MFCD00792441 CAS-Nummer: 700-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...