5-Brom-2-nitrophenol

Lieferant: COMBI-BLOCKS

AN-2554-5G AN-2554-1G
COBBAN-2554-5GEA 0 EUR
COBBAN-2554-5G COBBAN-2554-1G
5-Brom-2-nitrophenol
5-Brom-2-nitrophenol
Formel: C₆H₄BrNO₃
MDL: MFCD03095027
CAS-Nummer: 27684-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR