3-Brom-5-(trifluormethoxy)phenol
Lieferant: COMBI-BLOCKS
Synonyme:
1-Brom-3-hydroxy-5-(trifluormethoxy)benzol
OT-0763-1G
OT-0763-5G
COBBOT-0763-1GEA
623
EUR
COBBOT-0763-1G
COBBOT-0763-5G
3-Brom-5-(trifluormethoxy)phenol
3-Brom-5-(trifluormethoxy)phenol
Formel:
C₇H₄BrF₃O₂ Molecular Weight: 257.01 g/mol |
MDL:
MFCD15142929 CAS-Nummer: 1197239-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...