3-Brom-5-methoxyphenol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-3-hydroxy-5-methoxybenzol, 3-Brom-5-hydroxyanisol

OT-1238-1G OT-1238-5G OT-1238-250M
COBBOT-1238-1GEA 499 EUR
COBBOT-1238-1G COBBOT-1238-5G COBBOT-1238-250M
3-Brom-5-methoxyphenol
3-Brom-5-methoxyphenol
Formel: C₇H₇BrO₂
MDL: MFCD12405420
CAS-Nummer: 855400-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR