5-Brom-2-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromo-2-methoxy-1-nitrobenzene
AN-1462-5G
AN-1462-1G
COBBAN-1462-5GEA
359
EUR
COBBAN-1462-5G
COBBAN-1462-1G
5-Brom-2-nitroanisol
5-Brom-2-nitroanisol
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,03 g/mol |
MDL:
MFCD09966092 CAS-Nummer: 103966-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...